Assay Details
Target Gene Details
Entrez Gene ID: | 196549 |
Gene Name: | eukaryotic translation elongation factor 1 delta pseudogene 3 |
Gene Aliases: |
- |
Location: |
Chr.13:31846783-31959584 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1DP3 | NR_027062.1 | |||
| DA657873.1 |

Genomic Map