Assay Details
Target Gene Details
Entrez Gene ID: | 2098 |
Gene Name: | esterase D |
Gene Aliases: |
FGH |
Location: |
Chr.13:46771256-46797681 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESD | NM_001984.1 | NP_001975.1 | ||
| XM_005266278.2 | XP_005266335.1 | |||
| XM_011534954.1 | XP_011533256.1 | |||
| XM_017020414.1 | XP_016875903.1 | |||
| AF112219.1 | AAC99788.1 | |||
| AK098540.1 | ||||
| BC001169.1 | AAH01169.1 | |||
| D28416.1 | BAA05782.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763019 | Chr.13:46774094 - 47549312 on Build GRCh38 | Loss |
|
| esv3892341 | Chr.13:46772158 - 46817818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map