Assay Details
Target Gene Details
Entrez Gene ID: | 540 |
Gene Name: | ATPase copper transporting beta |
Gene Aliases: |
PWD, WC1, WD, WND |
Location: |
Chr.13:51932669-52012130 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
| nsv561649 | Chr.13:51917057 - 51996592 on Build GRCh38 | Loss |
|
| dgv1652n100 | Chr.13:51747428 - 52069788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map