Assay Details
Target Gene Details
Entrez Gene ID: | 1602 |
Gene Name: | dachshund family transcription factor 1 |
Gene Aliases: |
DACH |
Location: |
Chr.13:71437966-71868068 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACH1 | NM_004392.6 | NP_004383.4 | ||
| NM_080759.5 | NP_542937.3 | |||
| NM_080760.5 | NP_542938.3 | |||
| XM_011534939.2 | XP_011533241.1 | |||
| XM_011534942.2 | XP_011533244.1 | |||
| XM_017020396.1 | XP_016875885.1 | |||
| AF102546.1 | AAF01351.1 | |||
| AF356492.1 | AAL08487.1 | |||
| AJ005670.1 | CAA06666.1 | |||
| AK001000.1 | ||||
| AL079278.1 | CAB45153.1 | |||
| BC021219.2 | AAH21219.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471153 | Chr.13:71435470 - 71473964 on Build GRCh38 | Loss |
|
| nsv1038176 | Chr.13:70898018 - 71772254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map