Assay Details
Target Gene Details
Entrez Gene ID: | 337867 |
Gene Name: | UBA domain containing 2 |
Gene Aliases: |
PHGDHL1 |
Location: |
Chr.13:99200425-99386499 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBAC2 | NM_001144072.1 | 1 | 169 | NP_001137544.1 |
| XM_011521082.2 | 1 | 169 | XP_011519384.1 | |
| AK290342.1 | 1 | 169 |
Target Gene Details
Entrez Gene ID: | 100289373 |
Gene Name: | UBAC2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:99196374-99200757 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBAC2-AS1 | NR_036531.1 | 1 | 165 | |
| NR_036532.1 | ||||
| AK097488.1 | 1 | 170 | ||
| DB032212.1 | 1 | 165 | ||
| DB446060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892433 | Chr.13:98843569 - 99243711 on Build GRCh38 | Gain |
|
| nsv1050094 | Chr.13:98974062 - 99323969 on Build GRCh38 | Gain |
|
| esv3582749 | Chr.13:99137911 - 99253291 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map