Assay Details
Target Gene Details
Entrez Gene ID: | 51761 |
Gene Name: | ATPase phospholipid transporting 8A2 |
Gene Aliases: |
ATP, ATPIB, CAMRQ4, IB, ML-1 |
Location: |
Chr.13:25372011-26025851 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP8A2 | NM_016529.5 | NP_057613.4 | ||
| XM_011535103.1 | XP_011533405.1 | |||
| XM_011535106.1 | XP_011533408.1 | |||
| XM_011535107.2 | XP_011533409.1 | |||
| XM_011535113.2 | XP_011533415.1 | |||
| XM_017020625.1 | XP_016876114.1 | |||
| XM_017020626.1 | XP_016876115.1 | |||
| AK127263.1 | BAC86905.1 | |||
| DA326706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582237 | Chr.13:25162386 - 25434284 on Build GRCh38 | Gain |
|
| nsv510596 | Chr.13:25343323 - 25376831 on Build GRCh38 | Deletion |
|
| esv2747161 | Chr.13:25031743 - 26209531 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map