Assay Details
Target Gene Details
Entrez Gene ID: | 23348 |
Gene Name: | dedicator of cytokinesis 9 |
Gene Aliases: |
ZIZ1, ZIZIMIN1 |
Location: |
Chr.13:98793487-99086406 on Build GRCh38 |
Assay Gene Location: | Within Intron 50 |
More Information
Additional Information:
For this assay, SNP(s) [rs117143284] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map