Assay Details
Target Gene Details
Entrez Gene ID: | 80821 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
PA-PLA1, PAPLA1, SPG28 |
Location: |
Chr.14:53036740-53153328 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDHD1 | NM_001160147.1 | 13 | 9347 | NP_001153619.1 |
| NM_001160148.1 | 13 | 9410 | NP_001153620.1 | |
| NM_030637.2 | 12 | 9326 | NP_085140.2 | |
| XM_011537188.2 | 15 | 9522 | XP_011535490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832797 | Chr.14:52932853 - 53113976 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map