Assay Details
Target Gene Details
Entrez Gene ID: | 122945 |
Gene Name: | NADP dependent oxidoreductase domain containing 1 |
Gene Aliases: |
C14orf148 |
Location: |
Chr.14:77394021-77426013 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOXRED1 | NM_001113475.2 | 5 | 1012 | NP_001106946.1 |
| XM_005267330.3 | 6 | 1637 | XP_005267387.1 | |
| XM_011536427.2 | 6 | 1650 | XP_011534729.1 | |
| XM_011536428.2 | 6 | 897 | XP_011534730.1 | |
| XM_011536429.2 | 4 | 925 | XP_011534731.1 | |
| XM_017020969.1 | XP_016876458.1 | |||
| AK057371.1 | 4 | 1601 | ||
| BC066965.1 | 5 | 992 | AAH66965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1362 | Chr.14:77386859 - 77432262 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map