Assay Details
Target Gene Details
Entrez Gene ID: | 64067 |
Gene Name: | neuronal PAS domain protein 3 |
Gene Aliases: |
MOP6, PASD6, bHLHe12 |
Location: |
Chr.14:32934785-33804176 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121873 | Chr.14:32938694 - 32939394 on Build GRCh38 | Deletion |
|
| nsv1069789 | Chr.14:32938593 - 32939794 on Build GRCh38 | Deletion |
|
| nsv1126362 | Chr.14:32938394 - 32939794 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map