Assay Details
Target Gene Details
Entrez Gene ID: | 83694 |
Gene Name: | ribosomal protein S6 kinase like 1 |
Gene Aliases: |
RSKL2 |
Location: |
Chr.14:74903954-74923396 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
More Information
Set Membership: |
|

Genomic Map