Assay Details
Target Gene Details
Entrez Gene ID: | 161159 |
Gene Name: | ACTN1 antisense RNA 1 |
Gene Aliases: |
C14orf84 |
Location: |
Chr.14:68979682-68987463 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACTN1-AS1 | NR_073422.1 | |||
DB029126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832823 | Chr.14:68922136 - 69100921 on Build GRCh38 | Loss |
![]() ![]() ![]() |
