Assay Details
Target Gene Details
Entrez Gene ID: | 85439 |
Gene Name: | stonin 2 |
Gene Aliases: |
STN2, STNB, STNB2 |
Location: |
Chr.14:81260650-81427404 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STON2 | NM_001256430.1 | 8 | 3704 | NP_001243359.1 |
| NM_033104.3 | 6 | 12338 | NP_149095.2 | |
| XM_006720279.1 | 6 | 10055 | XP_006720342.1 | |
| XM_011537240.1 | 7 | 3660 | XP_011535542.1 | |
| XM_011537242.1 | 7 | 10226 | XP_011535544.1 | |
| AB208948.1 | 8 | 3704 | BAD92185.1 | |
| AK094799.1 | 3 | 1904 | ||
| BX648249.1 | 1 | 6022 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832841 | Chr.14:81121978 - 81277577 on Build GRCh38 | Gain |
|
| esv3635076 | Chr.14:80715257 - 81672373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map