Assay Details
Target Gene Details
Entrez Gene ID: | 12 |
Gene Name: | serpin family A member 3 |
Gene Aliases: |
AACT, ACT, GIG24, GIG25 |
Location: |
Chr.14:94612377-94624053 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA3 | NM_001085.4 | NP_001076.2 | ||
| AB209060.1 | BAD92297.1 | |||
| AF089747.1 | AAD08810.1 | |||
| AK093049.1 | ||||
| AK123091.1 | ||||
| AY513275.1 | AAT08028.1 | |||
| AY513276.1 | AAT08029.1 | |||
| BC003559.1 | ||||
| BC010530.1 | AAH10530.1 | |||
| BC034554.1 | AAH34554.1 | |||
| BC070265.1 | AAH70265.1 | |||
| J05176.1 | AAA51560.1 | |||
| K01500.1 | AAA51543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
|
| nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain |
|
| nsv832865 | Chr.14:94499457 - 94667894 on Build GRCh38 | Gain |
|
| esv3635384 | Chr.14:94569665 - 94707857 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map