Assay Details
Target Gene Details
Entrez Gene ID: | 5890 |
Gene Name: | RAD51 paralog B |
Gene Aliases: |
R51H2, RAD51L1, REC2 |
Location: |
Chr.14:67819779-68683118 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054697 | Chr.14:67735287 - 67879265 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map