Assay Details
Target Gene Details
Entrez Gene ID: | 89874 |
Gene Name: | solute carrier family 25 member 21 |
Gene Aliases: |
ODC, ODC1 |
Location: |
Chr.14:36677921-37172660 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A21 | NM_001171170.1 | NP_001164641.1 | ||
| NM_030631.3 | NP_085134.1 | |||
| XM_011537287.2 | XP_011535589.1 | |||
| XM_011537288.2 | XP_011535590.1 | |||
| XM_011537289.2 | XP_011535591.1 | |||
| AJ278148.1 | CAC27562.1 | |||
| AK289575.1 | ||||
| BC101521.1 | AAI01522.1 | |||
| BC113365.1 |
More Information
Set Membership: |
|

Genomic Map