Assay Details
Target Gene Details
Entrez Gene ID: | 117153 |
Gene Name: | melanoma inhibitory activity 2 |
Gene Aliases: |
- |
Location: |
Chr.14:39233921-39253371 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIA2 | NM_054024.3 | NP_473365.3 | ||
| AF390175.2 | AAL26990.2 | |||
| AK314069.1 | ||||
| BC035981.1 | AAH35981.1 | |||
| BC130537.1 | ||||
| BC130563.1 | ||||
| BX495701.1 | ||||
| BX648228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053133 | Chr.14:39188331 - 39301525 on Build GRCh38 | Gain |
|
| esv3584293 | Chr.14:39191534 - 39339749 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map