Assay Details
Target Gene Details
Entrez Gene ID: | 3183 |
Gene Name: | heterogeneous nuclear ribonucleoprotein C (C1/C2) |
Gene Aliases: |
C1, C2, HNRNP, HNRPC, SNRPC |
Location: |
Chr.14:21209136-21269479 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map