Assay Details
Target Gene Details
Entrez Gene ID: | 64067 |
Gene Name: | neuronal PAS domain protein 3 |
Gene Aliases: |
MOP6, PASD6, bHLHe12 |
Location: |
Chr.14:32934785-33804176 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPAS3 | XM_005267991.3 | XP_005268048.2 | ||
| XM_005267992.3 | XP_005268049.2 | |||
| XM_011537069.2 | XP_011535371.2 | |||
| XM_011537071.2 | XP_011535373.2 | |||
| XM_017021582.1 | XP_016877071.1 | |||
| XM_017021583.1 | XP_016877072.1 | |||
| XM_017021584.1 | XP_016877073.1 | |||
| XM_017021585.1 | XP_016877074.1 |
More Information
Set Membership: |
|

Genomic Map