Assay Details
Target Gene Details
Entrez Gene ID: | 753 |
Gene Name: | low density lipoprotein receptor class A domain containing 4 |
Gene Aliases: |
C18orf1 |
Location: |
Chr.18:13217691-13652754 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LDLRAD4 | NM_181482.4 | NP_852147.1 | ||
XM_005258140.1 | XP_005258197.1 | |||
XM_006722353.1 | XP_006722416.1 | |||
XM_011525737.1 | XP_011524039.1 | |||
XM_011525738.1 | XP_011524040.1 | |||
XM_017025961.1 | XP_016881450.1 | |||
XM_017025962.1 | XP_016881451.1 | |||
XM_017025963.1 | XP_016881452.1 | |||
XM_017025964.1 | XP_016881453.1 | |||
XM_017025965.1 | XP_016881454.1 | |||
XM_017025966.1 | XP_016881455.1 | |||
XM_017025967.1 | XP_016881456.1 | |||
XM_017025968.1 | XP_016881457.1 | |||
BC030199.1 | AAH30199.1 | |||
DB107684.1 | ||||
DB272592.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
