Assay Details
Target Gene Details
Entrez Gene ID: | 9745 |
Gene Name: | zinc finger protein 536 |
Gene Aliases: |
- |
Location: |
Chr.19:30224236-30713538 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF536 | XM_011527554.2 | 1 | 1035 | XP_011525856.1 |
| XM_011527555.2 | 1 | 1035 | XP_011525857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953271 | Chr.19:30222894 - 30229493 on Build GRCh38 | Deletion |
|
| nsv2458 | Chr.19:30195865 - 30240147 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map