Assay Details
Target Gene Details
Entrez Gene ID: | 199731 |
Gene Name: | cell adhesion molecule 4 |
Gene Aliases: |
IGSF4C, NECL4, Necl-4, TSLL2, synCAM4 |
Location: |
Chr.19:43622368-43641984 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CADM4 | XM_017026452.1 | XP_016881941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579852 | Chr.19:43638619 - 43645297 on Build GRCh38 | Loss |
|
| nsv833839 | Chr.19:43595661 - 43768147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map