Assay Details
Target Gene Details
Entrez Gene ID: | 10780 |
Gene Name: | zinc finger protein 234 |
Gene Aliases: |
HZF4, ZNF269 |
Location: |
Chr.19:44141530-44160309 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF234 | NM_001144824.1 | NP_001138296.1 | ||
| NM_006630.2 | NP_006621.1 | |||
| XM_006722974.3 | XP_006723037.1 | |||
| XM_017026149.1 | XP_016881638.1 | |||
| XM_017026150.1 | XP_016881639.1 | |||
| XM_017026151.1 | XP_016881640.1 | |||
| XM_017026152.1 | XP_016881641.1 | |||
| XM_017026153.1 | XP_016881642.1 | |||
| AK289843.1 | ||||
| AK314064.1 | ||||
| BC007303.1 | AAH07303.1 | |||
| X78927.1 | CAA55527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833840 | Chr.19:44016468 - 44211607 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75486190] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map