Assay Details
Target Gene Details
Entrez Gene ID: | 7730 |
Gene Name: | zinc finger protein 177 |
Gene Aliases: |
PIGX |
Location: |
Chr.19:9363020-9382617 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF177 | NM_001172651.1 | NP_001166122.1 | ||
| NM_003451.2 | NP_003442.2 | |||
| U37263.1 | AAB09749.1 |
Target Gene Details
Entrez Gene ID: | 100529215 |
Gene Name: | ZNF559-ZNF177 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:9324226-9382617 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF559-ZNF177 | NM_001172650.2 | NP_001166121.1 | ||
| NM_001202425.1 | NP_001189354.1 | |||
| U37251.1 | AAB09748.1 |
More Information
Set Membership: |
|

Genomic Map