Assay Details
Target Gene Details
Entrez Gene ID: | 10780 |
Gene Name: | zinc finger protein 234 |
Gene Aliases: |
HZF4, ZNF269 |
Location: |
Chr.19:44141530-44160309 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF234 | NM_001144824.1 | 6 | 3190 | NP_001138296.1 |
| NM_006630.2 | 6 | 3308 | NP_006621.1 | |
| XM_006722974.3 | XP_006723037.1 | |||
| XM_017026149.1 | XP_016881638.1 | |||
| XM_017026150.1 | XP_016881639.1 | |||
| XM_017026151.1 | XP_016881640.1 | |||
| XM_017026152.1 | XP_016881641.1 | |||
| XM_017026153.1 | XP_016881642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833840 | Chr.19:44016468 - 44211607 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map