Assay Details
Target Gene Details
Entrez Gene ID: | 126208 |
Gene Name: | zinc finger protein 787 |
Gene Aliases: |
TIP20 |
Location: |
Chr.19:56087363-56121373 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF787 | NM_001002836.3 | NP_001002836.2 | ||
| XM_011526445.2 | XP_011524747.1 | |||
| AF000560.1 | AAB58413.1 | |||
| BC077728.1 | AAH77728.1 | |||
| HY059733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833883 | Chr.19:56001235 - 56128720 on Build GRCh38 | Loss |
|
| dgv207n21 | Chr.19:56080920 - 56175584 on Build GRCh38 | Loss |
|
| dgv3681n100 | Chr.19:55906849 - 56338089 on Build GRCh38 | Gain |
|
| dgv206n21 | Chr.19:56026162 - 56195679 on Build GRCh38 | Loss |
|
| nsv580354 | Chr.19:55994319 - 56212123 on Build GRCh38 | Gain |
|
| nsv1057820 | Chr.19:56044576 - 56208893 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73059933,rs73934668] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map