Assay Details
Target Gene Details
Entrez Gene ID: | 84449 |
Gene Name: | zinc finger protein 333 |
Gene Aliases: |
- |
Location: |
Chr.19:14689787-14733746 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map