Assay Details
Target Gene Details
Entrez Gene ID: | 58513 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 like 1 |
Gene Aliases: |
EPS15R |
Location: |
Chr.19:16355244-16472012 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS15L1 | NM_001258374.1 | NP_001245303.1 | ||
| NR_047665.1 | ||||
| XM_017027086.1 | XP_016882575.1 | |||
| XM_017027087.1 | XP_016882576.1 | |||
| XM_017027088.1 | XP_016882577.1 | |||
| XM_017027089.1 | XP_016882578.1 | |||
| XM_017027090.1 | XP_016882579.1 | |||
| XM_017027091.1 | XP_016882580.1 | |||
| AK296473.1 | ||||
| AL110270.1 | CAH10724.1 | |||
| CA311813.1 |
More Information
Set Membership: |
|

Genomic Map