Assay Details
Target Gene Details
Entrez Gene ID: | 100533990 |
Gene Name: | APOC4-APOC2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.19:44942238-44949565 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOC4-APOC2 | NR_037932.1 | 3 | 867 | |
AK126257.1 | 4 | 801 |
