Assay Details
Target Gene Details
Entrez Gene ID: | 27113 |
Gene Name: | BCL2 binding component 3 |
Gene Aliases: |
JFY-1, JFY1, PUMA |
Location: |
Chr.19:47220822-47232998 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBC3 | NM_001127240.2 | NP_001120712.1 | ||
| NM_001127241.2 | NP_001120713.1 | |||
| NM_001127242.2 | NP_001120714.1 | |||
| XM_006723141.3 | XP_006723204.1 | |||
| AF354654.1 | AAK39542.1 | |||
| AF354655.1 | AAK39543.1 | |||
| AF354656.1 | AAK39544.1 | |||
| AK298837.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819260 | Chr.19:47228966 - 47232500 on Build GRCh38 | Loss |
|
| nsv833852 | Chr.19:47134590 - 47307834 on Build GRCh38 | Loss |
|
| esv26583 | Chr.19:47229711 - 47232430 on Build GRCh38 | Loss |
|
| nsv1067417 | Chr.19:47103045 - 47309908 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map