Assay Details
Target Gene Details
Entrez Gene ID: | 58513 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 like 1 |
Gene Aliases: |
EPS15R |
Location: |
Chr.19:16355244-16472012 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS15L1 | NM_001258374.1 | NP_001245303.1 | ||
| NM_001258375.1 | NP_001245304.1 | |||
| NM_021235.2 | NP_067058.1 | |||
| NR_047665.1 | ||||
| NR_047666.1 | ||||
| XM_017027086.1 | XP_016882575.1 | |||
| XM_017027087.1 | XP_016882576.1 | |||
| XM_017027088.1 | XP_016882577.1 | |||
| XM_017027089.1 | XP_016882578.1 | |||
| XM_017027090.1 | XP_016882579.1 | |||
| XM_017027091.1 | XP_016882580.1 | |||
| AB015346.1 | BAA88118.1 | |||
| AF110265.1 | AAF21930.1 | |||
| AK024166.1 | ||||
| AK025770.1 | ||||
| AK291359.1 | ||||
| AK296473.1 | ||||
| AK297428.1 | ||||
| AK299001.1 | ||||
| AL110270.1 | CAH10724.1 | |||
| BC131590.1 | ||||
| BU075732.1 |
More Information
Set Membership: |
|

Genomic Map