Assay Details
Target Gene Details
Entrez Gene ID: | 11136 |
Gene Name: | solute carrier family 7 member 9 |
Gene Aliases: |
BAT1, CSNU3 |
Location: |
Chr.19:32830511-32870958 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A9 | NM_001126335.1 | NP_001119807.1 | ||
| NM_001243036.1 | NP_001229965.1 | |||
| NM_014270.4 | NP_055085.1 | |||
| XM_006722992.1 | XP_006723055.1 | |||
| XM_011526402.2 | XP_011524704.1 | |||
| XM_017026230.1 | XP_016881719.1 | |||
| AB033548.1 | BAB16840.1 | |||
| AF141289.1 | AAD55898.1 | |||
| AJ249199.1 | CAB54003.1 | |||
| AK026446.1 | ||||
| AK223147.1 | BAD96867.1 | |||
| AK313708.1 | ||||
| AL365340.1 | ||||
| AL365341.1 | ||||
| AY170373.1 | ||||
| BC017962.1 | AAH17962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062926 | Chr.19:32828718 - 32904288 on Build GRCh38 | Loss |
|
| nsv516922 | Chr.19:32844072 - 32883216 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map