Assay Details
Target Gene Details
Entrez Gene ID: | 27335 |
Gene Name: | eukaryotic translation initiation factor 3 subunit K |
Gene Aliases: |
ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001 |
Location: |
Chr.19:38619072-38636959 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3K | NM_001300992.1 | NP_001287921.1 | ||
| NM_001308393.1 | NP_001295322.1 | |||
| NM_013234.3 | NP_037366.1 | |||
| AB019392.1 | BAA76626.1 | |||
| AF077051.1 | AAD27784.1 | |||
| AF085358.1 | AAD40193.1 | |||
| AF109355.1 | AAQ13503.1 | |||
| AF315506.1 | AAK01365.1 | |||
| AK026531.1 | ||||
| AK289554.1 | ||||
| AK298635.1 | ||||
| AK301035.1 | ||||
| AK316344.1 | ||||
| AY245432.1 | AAP22070.1 | |||
| BC001031.2 | AAH01031.1 | |||
| BC007559.1 | AAH07559.1 | |||
| BF965565.1 | ||||
| BG470776.1 | ||||
| BG488547.1 | ||||
| BG943109.1 | ||||
| BI116511.1 | ||||
| JX870647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833827 | Chr.19:38502414 - 38693907 on Build GRCh38 | Gain |
|
| nsv833828 | Chr.19:38594119 - 38767339 on Build GRCh38 | Loss |
|
| nsv1057901 | Chr.19:38621846 - 38683692 on Build GRCh38 | Gain |
|
| nsv953298 | Chr.19:38604961 - 38649760 on Build GRCh38 | Deletion |
|
| nsv470140 | Chr.19:38349499 - 38628854 on Build GRCh38 | Gain |
|
| nsv833826 | Chr.19:38454466 - 38693907 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map