Assay Details
Target Gene Details
Entrez Gene ID: | 64763 |
Gene Name: | zinc finger protein 574 |
Gene Aliases: |
FP972 |
Location: |
Chr.19:42068477-42081567 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF574 | XM_011527228.2 | XP_011525530.1 | ||
| XM_017027149.1 | XP_016882638.1 | |||
| AK131369.1 | BAD18520.1 | |||
| AK131418.1 | BAD18565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143455202] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map