Assay Details
Target Gene Details
Entrez Gene ID: | 90273 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 21 |
Gene Aliases: |
CEACAM3, R29124_1 |
Location: |
Chr.19:41549518-41586844 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM21 | NM_001288773.2 | NP_001275702.2 | ||
XM_011527450.1 | XP_011525752.1 | |||
XM_011527452.1 | XP_011525754.1 | |||
XM_017027433.1 | XP_016882922.1 | |||
XM_017027434.1 | XP_016882923.1 | |||
XM_017027435.1 | XP_016882924.1 | |||
XM_017027436.1 | XP_016882925.1 | |||
AK023602.1 | ||||
BI913326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519408 | Chr.19:41559909 - 41667978 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv3549n100 | Chr.19:41550966 - 41671232 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
