Assay Details
Target Gene Details
Entrez Gene ID: | 23309 |
Gene Name: | SIN3 transcription regulator family member B |
Gene Aliases: |
- |
Location: |
Chr.19:16829387-16880355 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIN3B | NM_001297595.1 | NP_001284524.1 | ||
| NM_015260.3 | NP_056075.1 | |||
| AB014600.2 | ||||
| AY706204.1 | AAU01916.1 | |||
| BC025026.1 | AAH25026.1 | |||
| BC063531.1 | AAH63531.1 | |||
| BC110821.1 | AAI10822.1 |
More Information
Set Membership: |
|

Genomic Map