Assay Details
Target Gene Details
Entrez Gene ID: | 2901 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 5 |
Gene Aliases: |
EAA2, GRIK2, GluK5, KA2 |
Location: |
Chr.19:41998316-42071804 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK5 | NM_001301030.1 | NP_001287959.1 | ||
| NM_002088.4 | NP_002079.3 | |||
| XM_005258821.3 | XP_005258878.1 | |||
| XM_011526862.2 | XP_011525164.1 | |||
| XM_011526863.2 | XP_011525165.1 | |||
| XM_011526865.1 | XP_011525167.1 | |||
| XM_011526867.1 | XP_011525169.1 | |||
| XM_011526868.1 | XP_011525170.1 | |||
| XM_017026713.1 | XP_016882202.1 | |||
| AJ249209.1 | CAC80547.1 | |||
| S40369.1 | AAB22591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map