Assay Details
Target Gene Details
Entrez Gene ID: | 9745 |
Gene Name: | zinc finger protein 536 |
Gene Aliases: |
- |
Location: |
Chr.19:30224236-30713538 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF536 | XM_011527554.2 | XP_011525856.1 | ||
| XM_011527555.2 | XP_011525857.1 | |||
| XM_011527558.2 | XP_011525860.1 | |||
| XM_017027527.1 | XP_016883016.1 | |||
| XM_017027528.1 | XP_016883017.1 | |||
| XM_017027529.1 | XP_016883018.1 | |||
| XM_017027530.1 | XP_016883019.1 | |||
| XM_017027531.1 | XP_016883020.1 | |||
| XM_017027532.1 | XP_016883021.1 | |||
| XM_017027533.1 | XP_016883022.1 | |||
| XM_017027534.1 | XP_016883023.1 | |||
| XM_017027535.1 | XP_016883024.1 | |||
| XM_017027536.1 | XP_016883025.1 | |||
| XM_017027537.1 | XP_016883026.1 | |||
| XM_017027538.1 | XP_016883027.1 | |||
| XM_017027539.1 | XP_016883028.1 | |||
| XM_017027540.1 | XP_016883029.1 | |||
| XM_017027542.1 | XP_016883031.1 |
More Information
Set Membership: |
|

Genomic Map