Assay Details
Target Gene Details
Entrez Gene ID: | 57794 |
Gene Name: | SURP and G-patch domain containing 1 |
Gene Aliases: |
F23858, RBP, SF4 |
Location: |
Chr.19:19276513-19320512 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGP1 | NM_172231.3 | NP_757386.2 | ||
| XM_005260002.2 | XP_005260059.1 | |||
| XM_011528155.2 | XP_011526457.1 | |||
| AF521128.1 | AAN77123.1 | |||
| AK096152.1 | ||||
| AK292085.1 | ||||
| AK294618.1 | ||||
| AL137286.1 | CAB70678.1 | |||
| AL713757.1 | CAD28528.1 | |||
| AY072916.1 | AAL68960.1 | |||
| AY072917.1 | AAL68961.1 | |||
| BC063784.1 | AAH63784.1 | |||
| BC142988.1 |
More Information
Set Membership: |
|

Genomic Map