Assay Details
Target Gene Details
Entrez Gene ID: | 79085 |
Gene Name: | solute carrier family 25 member 23 |
Gene Aliases: |
APC2, MCSC2, SCaMC-3 |
Location: |
Chr.19:6436081-6460221 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A23 | XM_011528274.1 | XP_011526576.1 | ||
| XM_011528275.1 | XP_011526577.1 | |||
| XM_011528276.1 | XP_011526578.1 | |||
| XM_011528277.1 | XP_011526579.1 | |||
| XM_011528278.1 | XP_011526580.1 | |||
| XM_011528279.1 | XP_011526581.1 | |||
| XM_011528280.1 | XP_011526582.1 | |||
| XM_011528282.1 | XP_011526584.1 | |||
| XM_011528283.1 | XP_011526585.1 | |||
| XM_011528284.1 | XP_011526586.1 | |||
| XM_017027285.1 | XP_016882774.1 | |||
| XM_017027286.1 | XP_016882775.1 | |||
| XM_017027287.1 | XP_016882776.1 | |||
| XM_017027288.1 | XP_016882777.1 | |||
| XM_017027292.1 | XP_016882781.1 | |||
| AJ879080.1 | CAI51682.1 | |||
| AJ879081.1 | CAI51683.1 | |||
| AJ879082.1 | CAI51684.1 | |||
| AJ879083.1 | CAI51685.1 | |||
| BC001656.1 | AAH01656.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map