Assay Details
Target Gene Details
Entrez Gene ID: | 2109 |
Gene Name: | electron transfer flavoprotein beta subunit |
Gene Aliases: |
FP585, MADD |
Location: |
Chr.19:51345155-51366418 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETFB | NM_001985.2 | NP_001976.1 | ||
| AK291881.1 | ||||
| BC093961.1 | AAH93961.1 | |||
| BC093963.1 | AAH93963.1 | |||
| BG399783.1 | ||||
| CR456827.1 | CAG33108.1 | |||
| X71129.1 | CAA50441.1 |
More Information
Set Membership: |
|

Genomic Map