Assay Details
Target Gene Details
Entrez Gene ID: | 4793 |
Gene Name: | NFKB inhibitor beta |
Gene Aliases: |
IKBB, TRIP9 |
Location: |
Chr.19:38899503-38908894 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKBIB | NM_001243116.1 | NP_001230045.1 | ||
| NM_002503.4 | NP_002494.2 | |||
| NR_040515.1 | ||||
| XM_006723226.2 | XP_006723289.2 | |||
| XM_006723227.3 | XP_006723290.3 | |||
| AB209784.1 | BAD93021.1 | |||
| AB362577.1 | ||||
| AK057862.1 | BAB71599.1 | |||
| AK290569.1 | ||||
| BC015528.1 | AAH15528.1 | |||
| BI520169.1 | ||||
| BM008311.1 | ||||
| BM449613.1 | ||||
| BQ440907.1 | ||||
| BT006743.1 | AAP35389.1 | |||
| DA633677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160616 | Chr.19:38839331 - 38903590 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map