Assay Details
Target Gene Details
Entrez Gene ID: | 60680 |
Gene Name: | CUGBP, Elav-like family member 5 |
Gene Aliases: |
BRUNOL-5, BRUNOL5, CELF-5 |
Location: |
Chr.19:3224676-3297076 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF5 | NM_001172673.1 | NP_001166144.1 | ||
| NM_021938.3 | NP_068757.2 | |||
| NR_033342.1 | ||||
| XM_006722832.1 | XP_006722895.1 | |||
| XM_006722833.1 | XP_006722896.1 | |||
| XM_006722837.3 | XP_006722900.1 | |||
| AB209067.1 | BAD92304.1 | |||
| AF329266.1 | AAK07476.1 | |||
| AK294109.1 | ||||
| BC028101.1 | AAH28101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509716 | Chr.19:3077662 - 3262653 on Build GRCh38 | Insertion |
|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map