Assay Details
Target Gene Details
Entrez Gene ID: | 7040 |
Gene Name: | transforming growth factor beta 1 |
Gene Aliases: |
CED, DPD1, LAP, TGFB, TGFbeta |
Location: |
Chr.19:41330531-41353933 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB1 | NM_000660.5 | NP_000651.3 | ||
| XM_011527242.1 | XP_011525544.1 | |||
| AK291907.1 | ||||
| AK307742.1 | ||||
| AY820829.1 | AAV71148.1 | |||
| BC000125.1 | AAH00125.1 | |||
| BC001180.1 | AAH01180.1 | |||
| BC022242.1 | AAH22242.1 | |||
| BT007245.1 | AAP35909.1 | |||
| X02812.1 | CAA26580.1 |
More Information
Set Membership: |
|

Genomic Map