Assay Details
Target Gene Details
Entrez Gene ID: | 11140 |
Gene Name: | cell division cycle 37 |
Gene Aliases: |
P50CDC37 |
Location: |
Chr.19:10391128-10403595 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC37 | NM_007065.3 | NP_008996.1 | ||
| XM_011527652.2 | XP_011525954.1 | |||
| BC000083.2 | AAH00083.1 | |||
| BC008793.2 | AAH08793.1 | |||
| BC127212.1 | ||||
| BC127213.1 | ||||
| BC141928.1 | ||||
| BC141929.1 | ||||
| BT006796.1 | AAP35442.1 | |||
| CR542249.1 | CAG47045.1 | |||
| U43077.1 | AAB63979.1 | |||
| U63131.1 | AAB04798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518008 | Chr.19:10338682 - 10426592 on Build GRCh38 | Gain |
|
| esv24860 | Chr.19:10378463 - 10394341 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map