Assay Details
Target Gene Details
Entrez Gene ID: | 2821 |
Gene Name: | glucose-6-phosphate isomerase |
Gene Aliases: |
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36 |
Location: |
Chr.19:34359592-34402413 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPI | NM_000175.3 | NP_000166.2 | ||
| NM_001184722.1 | NP_001171651.1 | |||
| NM_001289789.1 | NP_001276718.1 | |||
| NM_001289790.1 | NP_001276719.1 | |||
| XM_005258764.2 | XP_005258821.1 | |||
| XM_006723148.2 | XP_006723211.1 | |||
| XM_011526754.2 | XP_011525056.1 | |||
| AK025421.1 | ||||
| AK129884.1 | ||||
| AK130102.1 | ||||
| AK293446.1 | ||||
| AK294396.1 | ||||
| AK301103.1 | ||||
| BC004982.1 | AAH04982.1 | |||
| K03515.1 | AAA36368.1 | |||
| X16539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644206 | Chr.19:34296741 - 34419464 on Build GRCh38 | Loss |
|
| nsv509738 | Chr.19:34351799 - 34412408 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map