Assay Details
Target Gene Details
Entrez Gene ID: | 5655 |
Gene Name: | kallikrein related peptidase 10 |
Gene Aliases: |
NES1, PRSSL1 |
Location: |
Chr.19:51012744-51020175 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK10 | NM_001077500.1 | 6 | 1615 | NP_001070968.1 |
| NM_002776.4 | 6 | 1714 | NP_002767.2 | |
| NM_145888.2 | 6 | 1576 | NP_665895.1 | |
| AK026045.1 | 1 | 219 |
More Information
Set Membership: |
|

Genomic Map