Assay Details
Target Gene Details
Entrez Gene ID: | 593 |
Gene Name: | branched chain keto acid dehydrogenase E1, alpha polypeptide |
Gene Aliases: |
BCKDE1A, MSU, MSUD1, OVD1A |
Location: |
Chr.19:41397789-41425005 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCKDHA | NM_000709.3 | NP_000700.1 | ||
| NM_001164783.1 | NP_001158255.1 | |||
| AB209828.1 | BAD93065.1 | |||
| AK298188.1 | ||||
| AK313300.1 | ||||
| AL833143.1 | CAH56215.1 | |||
| BC007878.2 | AAH07878.1 | |||
| BC008933.2 | AAH08933.1 | |||
| BC023983.1 | AAH23983.1 | |||
| BG742673.1 | ||||
| BI910860.1 | ||||
| J04474.1 | AAB59549.1 | |||
| M22221.1 | AAA35590.1 | |||
| Z14093.1 | CAA78475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644379 | Chr.19:41417796 - 41423703 on Build GRCh38 | Loss |
|
| nsv953578 | Chr.19:41421196 - 41428795 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map