Assay Details
Target Gene Details
Entrez Gene ID: | 374887 |
Gene Name: | YjeF N-terminal domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.19:19528861-19537584 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YJEFN3 | NM_001190328.1 | NP_001177257.1 | ||
| NM_198537.3 | NP_940939.2 | |||
| XM_005259903.4 | XP_005259960.1 | |||
| XM_011527997.1 | XP_011526299.1 | |||
| XM_011527998.2 | XP_011526300.1 | |||
| XM_011527999.1 | XP_011526301.1 | |||
| XM_017026770.1 | XP_016882259.1 | |||
| XM_017026771.1 | XP_016882260.1 | |||
| AK094976.1 | 1 | 138 | ||
| AK126916.1 | 3 | 918 | BAC86749.1 | |
| BI520231.1 | ||||
| DQ778079.1 | ||||
| DQ778080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509723 | Chr.19:19517547 - 19595561 on Build GRCh38 | Insertion |
|
| nsv952426 | Chr.19:19525792 - 19601091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map