Assay Details
Target Gene Details
| Entrez Gene ID: | 2909 | 
| Gene Name: | Rho GTPase activating protein 35 | 
| Gene Aliases: | GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP | 
| Location: | Chr.19:46861002-47005077 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ARHGAP35 | XM_011526873.2 | XP_011525175.1 | ||
| XM_017026714.1 | XP_016882203.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3893224 | Chr.19:46783912 - 46911729 on Build GRCh38 | Gain |  SNAR-E  ARHGAP35  AP2S1  SLC1A5 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top